siRNA Details
| Gene Symbol: Sncb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080777.2 | NP_542955.2 | 2 | 255 | ||
| XM_006253594.2 | XP_006253656.1 | 3 | 456 | |||
| GenBank mRNA | BC087579.1 | - | 2 | 255 | 7314241 | |
| D17764.1 | - | 2 | 111 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sncb| Gene Symbol: Sncb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080777.2 | NP_542955.2 | 2 | 255 | ||
| XM_006253594.2 | XP_006253656.1 | 3 | 456 | |||
| GenBank mRNA | BC087579.1 | - | 2 | 255 | 7314241 | |
| D17764.1 | - | 2 | 111 | |||