siRNA Details
| Gene Symbol: Rtn4r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053613.1 | NP_446065.1 | 2 | 338 | ||
| XM_008768839.1 | XP_008767061.1 | 2 | 871 | |||
| GenBank mRNA | AF462390.1 | - | 2 | 338 | ||
| AY028438.1 | - | 2 | 338 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rtn4r| Gene Symbol: Rtn4r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053613.1 | NP_446065.1 | 2 | 338 | ||
| XM_008768839.1 | XP_008767061.1 | 2 | 871 | |||
| GenBank mRNA | AF462390.1 | - | 2 | 338 | ||
| AY028438.1 | - | 2 | 338 | |||