siRNA Details
| Gene Symbol: Nup88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053616.1 | NP_446068.1 | 4 | 654 | ||
| GenBank mRNA | BC072524.1 | - | 4 | 648 | 7097375 | |
| U93692.1 | - | 4 | 654 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nup88| Gene Symbol: Nup88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053616.1 | NP_446068.1 | 4 | 654 | ||
| GenBank mRNA | BC072524.1 | - | 4 | 648 | 7097375 | |
| U93692.1 | - | 4 | 654 | |||