siRNA Details
| Gene Symbol: TAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003185.3 | NP_003176.2 | 6 | 1927 | ||
| GenBank mRNA | AK123937.1 | - | 2 | 776 | ||
| U75308.1 | - | 8 | 1513 | |||
| Y11354.1 | - | 6 | 1921 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAF4| Gene Symbol: TAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003185.3 | NP_003176.2 | 6 | 1927 | ||
| GenBank mRNA | AK123937.1 | - | 2 | 776 | ||
| U75308.1 | - | 8 | 1513 | |||
| Y11354.1 | - | 6 | 1921 | |||