siRNA Details
| Gene Symbol: Clec2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130402.2 | NP_569086.1 | 4, 5 | 583 | ||
| XM_008763309.1 | XP_008761531.1 | 5, 6 | 1080 | |||
| GenBank mRNA | AF321552.1 | - | 6, 7 | 769 | ||
| EF100688.1 | - | 4, 5 | 583 | |||

Clec2d
Genomic Map