siRNA Details
| Gene Symbol: Clec2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130402.2 | NP_569086.1 | 2 | 183 | ||
| XM_008763309.1 | XP_008761531.1 | 3 | 680 | |||
| GenBank mRNA | AF321552.1 | - | 4 | 369 | ||
| EF100688.1 | - | 2 | 183 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec2d| Gene Symbol: Clec2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130402.2 | NP_569086.1 | 2 | 183 | ||
| XM_008763309.1 | XP_008761531.1 | 3 | 680 | |||
| GenBank mRNA | AF321552.1 | - | 4 | 369 | ||
| EF100688.1 | - | 2 | 183 | |||