siRNA Details
| Gene Symbol: Gmfg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181091.2 | NP_851605.1 | 3, 4 | 213 | ||
| GenBank mRNA | AB007364.1 | - | 3, 4 | 213 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gmfg| Gene Symbol: Gmfg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181091.2 | NP_851605.1 | 3, 4 | 213 | ||
| GenBank mRNA | AB007364.1 | - | 3, 4 | 213 | ||