siRNA Details
| Gene Symbol: Gmfg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181091.2 | NP_851605.1 | 1 | 50 | ||
| XM_006228606.2 | XP_006228668.1 | 2 | 348 | |||
| XM_006228607.2 | XP_006228669.1 | 2 | 348 | |||
| XM_008759100.1 | XP_008757322.1 | 2 | 163 | |||
| GenBank mRNA | AB007364.1 | - | 1 | 50 | ||

Gmfg
Genomic Map