siRNA Details
| Gene Symbol: Maged2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271109.1 | NP_001258038.1 | 10 | 1315 | ||
| NM_001271110.1 | NP_001258039.1 | 10 | 1343 | |||
| NM_080479.2 | NP_536727.2 | 10 | 1315 | |||
| XM_006237924.2 | XP_006237986.1 | 10 | 1405 | |||
| GenBank mRNA | AJ293617.1 | - | 9 | 1256 | ||
| AY724507.1 | - | 10 | 1328 | |||
| BC086998.1 | - | 8 | 2272 | |||
| BC107467.1 | - | 10 | 1315 | 7459969 | ||
| FQ229095.1 | - | 10 | 1328 | |||

Maged2
Genomic Map