siRNA Details
| Gene Symbol: Pecr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133299.2 | NP_579833.1 | 7 | 912 | ||
| XM_008767180.1 | XP_008765402.1 | 7 | 978 | |||
| GenBank mRNA | AF021854.1 | - | 7 | 913 | ||
| AF099742.1 | - | 7 | 832 | |||
| BC060546.1 | - | 7 | 906 | 6889831 | ||
| FQ209650.1 | - | 4 | 389 | |||
| FQ210848.1 | - | 4 | 389 | |||
| FQ218655.1 | - | 4 | 389 | |||
| FQ218673.1 | - | 4 | 389 | |||
| FQ218683.1 | - | 4 | 389 | |||
| FQ218690.1 | - | 4 | 389 | |||
| FQ219508.1 | - | 4 | 389 | |||

Pecr
Genomic Map