siRNA Details
| Gene Symbol: Mrps7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100471.1 | NP_001093941.1 | 4 | 418 | ||
| GenBank mRNA | BC088232.1 | - | 4 | 407 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrps7| Gene Symbol: Mrps7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100471.1 | NP_001093941.1 | 4 | 418 | ||
| GenBank mRNA | BC088232.1 | - | 4 | 407 | ||