siRNA Details
| Gene Symbol: C5ar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053619.1 | NP_446071.1 | 2 | 571 | ||
| GenBank mRNA | AB003042.1 | - | 2 | 571 | ||
| AF090996.1 | - | 1 | 286 | |||
| BC078770.1 | - | 2 | 554 | 7121456 | ||
| Y09613.1 | - | 2 | 555 | |||
More Information
Set Membership: |
|

C5ar1
Genomic Map