siRNA Details
| Gene Symbol: Nr2f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080778.2 | NP_542956.1 | 3 | 1374 | ||
| XM_006229366.2 | XP_006229428.1 | 3 | 6261 | |||
| GenBank mRNA | AF003944.1 | - | 3 | 1375 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_183261, NM_009697, BC094360, AK032882, BC042484, and others.

Nr2f2
Genomic Map