siRNA Details
| Gene Symbol: Nr2f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080778.2 | NP_542956.1 | 3 | 1222 | ||
| XM_006229366.2 | XP_006229428.1 | 3 | 6109 | |||
| GenBank mRNA | AF003944.1 | - | 3 | 1223 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nr2f2| Gene Symbol: Nr2f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080778.2 | NP_542956.1 | 3 | 1222 | ||
| XM_006229366.2 | XP_006229428.1 | 3 | 6109 | |||
| GenBank mRNA | AF003944.1 | - | 3 | 1223 | ||