siRNA Details
| Gene Symbol: P2rx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080780.2 | NP_542958.2 | 1, 2 | 313 | ||
| XM_006246566.2 | XP_006246628.1 | 1, 2 | 393 | |||
| GenBank mRNA | X92069.1 | - | 1, 2 | 313 | ||
| X97328.1 | - | 1, 2 | 179 | |||

P2rx5
Genomic Map