siRNA Details
| Gene Symbol: Ppp1r14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130403.1 | NP_569087.1 | 4 | 514 | ||
| GenBank mRNA | AF352572.1 | - | 4 | 514 | ||
| AY050673.1 | - | 4 | 514 | |||
| FQ230520.1 | - | 4 | 506 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp1r14a| Gene Symbol: Ppp1r14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130403.1 | NP_569087.1 | 4 | 514 | ||
| GenBank mRNA | AF352572.1 | - | 4 | 514 | ||
| AY050673.1 | - | 4 | 514 | |||
| FQ230520.1 | - | 4 | 506 | |||