siRNA Details
| Gene Symbol: Ebna1bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008721.1 | NP_001008721.1 | 519 | |||
| GenBank mRNA | BC087738.1 | - | 519 | 7310934 | ||
| DQ480746.1 | - | 1508 | ||||
| FQ213511.1 | - | 540 | ||||
| FQ231568.1 | - | 541 | ||||
| Gene Symbol: LOC100912375 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750007.3 | XP_003750055.1 | 4, 5 | 539 | ||

Ebna1bp2
Genomic Map