siRNA Details
| Gene Symbol: Ctnnd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271502.1 | NP_001258431.1 | 9 | 1607 | ||
| XM_006232094.2 | XP_006232156.1 | 9 | 2584 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctnnd2| Gene Symbol: Ctnnd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271502.1 | NP_001258431.1 | 9 | 1607 | ||
| XM_006232094.2 | XP_006232156.1 | 9 | 2584 | |||