siRNA Details
Gene Symbol: Sec16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276417.1 | NP_001263346.1 | 2 | 3880 | ||
XM_006233662.2 | XP_006233724.1 | 2 | 3904 | |||
XM_006233663.2 | XP_006233725.1 | 2 | 3707 | |||
XM_006233664.2 | XP_006233726.1 | 2 | 4790 | |||
XM_006233666.2 | XP_006233728.1 | 3 | 3214 |
