siRNA Details
| Gene Symbol: St14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053635.2 | NP_446087.1 | 11 | 1402 | ||
| GenBank mRNA | AB037898.1 | - | 11 | 1276 | ||
| AB049189.1 | - | 11 | 1402 | |||
| BC097271.1 | - | 11 | 1307 | 7445508 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

St14| Gene Symbol: St14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053635.2 | NP_446087.1 | 11 | 1402 | ||
| GenBank mRNA | AB037898.1 | - | 11 | 1276 | ||
| AB049189.1 | - | 11 | 1402 | |||
| BC097271.1 | - | 11 | 1307 | 7445508 | ||