siRNA Details
| Gene Symbol: Ltc4s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053639.2 | NP_446091.2 | 1, 2 | 87 | ||
| XM_008767655.1 | XP_008765877.1 | 2, 3 | 1108 | |||
| GenBank mRNA | AB048790.2 | - | 1, 2 | 87 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ltc4s| Gene Symbol: Ltc4s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053639.2 | NP_446091.2 | 1, 2 | 87 | ||
| XM_008767655.1 | XP_008765877.1 | 2, 3 | 1108 | |||
| GenBank mRNA | AB048790.2 | - | 1, 2 | 87 | ||