siRNA Details
| Gene Symbol: Nkx2-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053651.1 | NP_446103.1 | 1 | 413 | ||
| GenBank mRNA | AF006664.1 | - | 1 | 413 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nkx2-5| Gene Symbol: Nkx2-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053651.1 | NP_446103.1 | 1 | 413 | ||
| GenBank mRNA | AF006664.1 | - | 1 | 413 | ||