siRNA Details
| Gene Symbol: Ccnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053662.3 | NP_446114.1 | 731 | |||
| XM_008761029.1 | XP_008759251.1 | 1807 | ||||
| GenBank mRNA | AF030091.3 | - | 731 | |||
| BC085686.1 | - | 785 | ||||
| BC128699.1 | - | 825 | ||||
| BC161817.1 | - | 677 | 9022416 | |||
| FQ213168.1 | - | 2 | 155 | |||
| FQ232436.1 | - | 768 | ||||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_019937, AK011629, AF467251, BC094383, BC117816, and others.

Ccnl1
Genomic Map