siRNA Details
| Gene Symbol: Akap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053665.1 | NP_446117.1 | 6 | 2274 | ||
| XM_006247067.2 | XP_006247129.1 | 7 | 2447 | |||
| XM_006247068.1 | XP_006247130.1 | 7 | 2443 | |||
| XM_008767930.1 | XP_008766152.1 | 7 | 2502 | |||
| GenBank mRNA | AF068202.1 | - | 6 | 2274 | ||
| BC081753.1 | - | 8 | 2526 | |||
| BC097298.1 | - | 9 | 2618 | |||
| BC129090.1 | - | 9 | 2597 | |||

Akap1
Genomic Map