siRNA Details
| Gene Symbol: Akap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053665.1 | NP_446117.1 | 4 | 1895 | ||
| XM_006247067.2 | XP_006247129.1 | 5 | 2068 | |||
| XM_006247068.1 | XP_006247130.1 | 5 | 2064 | |||
| XM_008767930.1 | XP_008766152.1 | 5 | 2123 | |||
| GenBank mRNA | AF068202.1 | - | 4 | 1895 | ||
| BC081753.1 | - | 6 | 2147 | |||
| BC097298.1 | - | 7 | 2239 | |||
| BC129090.1 | - | 7 | 2218 | |||

Akap1
Genomic Map