siRNA Details
| Gene Symbol: P3h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053667.1 | NP_446119.1 | 4 | 919 | ||
| GenBank mRNA | AF087433.1 | - | 4 | 919 | ||
| BC161838.1 | - | 4 | 953 | 9101602 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

P3h1| Gene Symbol: P3h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053667.1 | NP_446119.1 | 4 | 919 | ||
| GenBank mRNA | AF087433.1 | - | 4 | 919 | ||
| BC161838.1 | - | 4 | 953 | 9101602 | ||