siRNA Details
| Gene Symbol: Mpp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053668.1 | NP_446120.1 | 18, 19 | 1727 | ||
| XM_008767931.1 | XP_008766153.1 | 18, 19 | 1703 | |||
| XM_008767932.1 | XP_008766154.1 | 18, 19 | 1827 | |||
| GenBank mRNA | FQ211573.1 | - | 18, 19 | 1854 | ||

Mpp3
Genomic Map