siRNA Details
Gene Symbol: LOC102548101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762223.1 | XP_008760445.1 | 1 | 464 |
Gene Symbol: Tmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053671.2 | NP_446123.2 | 5 | 1681 | ||
XM_008763143.1 | XP_008761365.1 | 5 | 3203 | |||
GenBank mRNA | AF107843.1 | - | 5 | 1604 | ||
CK473511.1 | - | 4 | 533 |
