siRNA Details
| Gene Symbol: Phyh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053674.1 | NP_446126.1 | 2, 3 | 142 | ||
| GenBank mRNA | AF121345.1 | - | 2, 3 | 142 | ||
| BC086573.1 | - | 2, 3 | 137 | 7306754 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phyh| Gene Symbol: Phyh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053674.1 | NP_446126.1 | 2, 3 | 142 | ||
| GenBank mRNA | AF121345.1 | - | 2, 3 | 142 | ||
| BC086573.1 | - | 2, 3 | 137 | 7306754 | ||