siRNA Details
| Gene Symbol: Spata2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053675.2 | NP_446127.3 | 3 | 839 | ||
| XM_006235636.2 | XP_006235698.1 | 3 | 750 | |||
| GenBank mRNA | BC081752.1 | - | 3 | 839 | 7134513 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata2| Gene Symbol: Spata2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053675.2 | NP_446127.3 | 3 | 839 | ||
| XM_006235636.2 | XP_006235698.1 | 3 | 750 | |||
| GenBank mRNA | BC081752.1 | - | 3 | 839 | 7134513 | |