siRNA Details
| Gene Symbol: Spata2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053675.2 | NP_446127.3 | 3 | 1813 | ||
| XM_006235636.2 | XP_006235698.1 | 3 | 1724 | |||
| GenBank mRNA | AF123651.1 | - | 2 | 1614 | ||
| BC081752.1 | - | 3 | 1813 | 7134513 | ||

Spata2
Genomic Map