siRNA Details
| Gene Symbol: Chek2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053677.1 | NP_446129.1 | 10, 11 | 1197 | ||
| XM_006249518.2 | XP_006249580.1 | 11, 12 | 1311 | |||
| XM_006249519.2 | XP_006249581.1 | 10, 11 | 1190 | |||
| XM_006249520.2 | XP_006249582.1 | 10, 11 | 1247 | |||
| XM_006249521.2 | XP_006249583.1 | 10, 11 | 1131 | |||
| XM_008769308.1 | XP_008767530.1 | 10, 11 | 1190 | |||
| GenBank mRNA | AF134054.1 | - | 10, 11 | 1197 | ||

Chek2
Genomic Map