siRNA Details
| Gene Symbol: Rax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053678.1 | NP_446130.1 | 3 | 600 | ||
| GenBank mRNA | AF135839.1 | - | 3 | 600 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rax| Gene Symbol: Rax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053678.1 | NP_446130.1 | 3 | 600 | ||
| GenBank mRNA | AF135839.1 | - | 3 | 600 | ||