siRNA Details
| Gene Symbol: Dffa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053679.2 | NP_446131.2 | 4, 5 | 697 | ||
| XM_006239362.2 | XP_006239424.1 | 4, 5 | 718 | |||
| GenBank mRNA | AF136601.1 | - | 4, 5 | 682 | ||
| BC100067.1 | - | 4, 5 | 697 | 7389030 | ||

Dffa
Genomic Map