siRNA Details
| Gene Symbol: S100a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053681.1 | NP_446133.1 | 3 | 205 | ||
| XM_006232558.2 | XP_006232620.1 | 3 | 408 | |||
| XM_006232559.2 | XP_006232621.1 | 3 | 1162 | |||
| XM_006232560.2 | XP_006232622.1 | 3 | 1611 | |||
| XM_006232563.2 | XP_006232625.1 | 3 | 193 | |||
| XM_008761122.1 | XP_008759344.1 | 4 | 640 | |||
| XM_008761123.1 | XP_008759345.1 | 3 | 683 | |||
| GenBank mRNA | AF140231.1 | - | 3 | 205 | ||

S100a3
Genomic Map