siRNA Details
| Gene Symbol: Pde6h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053688.2 | NP_446140.1 | 2, 3 | 187 | ||
| XM_006237558.2 | XP_006237620.1 | 3, 4 | 1515 | |||
| GenBank mRNA | AF169390.1 | - | 2, 3 | 177 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pde6h| Gene Symbol: Pde6h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053688.2 | NP_446140.1 | 2, 3 | 187 | ||
| XM_006237558.2 | XP_006237620.1 | 3, 4 | 1515 | |||
| GenBank mRNA | AF169390.1 | - | 2, 3 | 177 | ||