siRNA Details
| Gene Symbol: Dmtf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053693.1 | NP_446145.1 | 3, 4 | 389 | ||
| XM_006235993.1 | XP_006236055.1 | 2, 3 | 241 | |||
| GenBank mRNA | BC081880.1 | - | 3, 4 | 389 | 7108159 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dmtf1| Gene Symbol: Dmtf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053693.1 | NP_446145.1 | 3, 4 | 389 | ||
| XM_006235993.1 | XP_006236055.1 | 2, 3 | 241 | |||
| GenBank mRNA | BC081880.1 | - | 3, 4 | 389 | 7108159 | |