siRNA Details
Gene Symbol: Wnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_575397.6 | XP_575397.1 | 1, 2 | 619 | ||
GenBank mRNA | AF352177.1 | - | 1, 2 | 157 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
