siRNA Details
Gene Symbol: Cited2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053698.2 | NP_446150.1 | 2 | 925 | ||
GenBank mRNA | AF361476.1 | - | 2 | 930 | ||
BC087005.1 | - | 2 | 925 | 7191504 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cited2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053698.2 | NP_446150.1 | 2 | 925 | ||
GenBank mRNA | AF361476.1 | - | 2 | 930 | ||
BC087005.1 | - | 2 | 925 | 7191504 |