siRNA Details
| Gene Symbol: Ccna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053702.3 | NP_446154.3 | 7 | 1209 | ||
| XM_006232266.2 | XP_006232328.1 | 8 | 1346 | |||
| XM_008760903.1 | XP_008759125.1 | 8 | 1341 | |||
| GenBank mRNA | AY623027.1 | - | 6 | 1183 | ||
| AY623028.1 | - | 7 | 1588 | |||
| BC127462.1 | - | 8 | 1302 | 8373429 | ||
| BF406951.1 | - | 4 | 384 | |||
| CV114678.1 | - | 4 | 476 | |||

Ccna2
Genomic Map