siRNA Details
| Gene Symbol: Ccna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053702.3 | NP_446154.3 | 2, 3 | 511 | ||
| GenBank mRNA | AF367448.1 | - | 2, 3 | 410 | ||
| AY623027.1 | - | 1, 2 | 485 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccna2| Gene Symbol: Ccna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053702.3 | NP_446154.3 | 2, 3 | 511 | ||
| GenBank mRNA | AF367448.1 | - | 2, 3 | 410 | ||
| AY623027.1 | - | 1, 2 | 485 | |||