siRNA Details
Gene Symbol: Map2k6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053703.3 | NP_446155.1 | 5 | 437 | ||
GenBank mRNA | AF369384.1 | - | 5 | 437 | ||
BC087004.1 | - | 5 | 537 | 7190965 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Map2k6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053703.3 | NP_446155.1 | 5 | 437 | ||
GenBank mRNA | AF369384.1 | - | 5 | 437 | ||
BC087004.1 | - | 5 | 537 | 7190965 |