siRNA Details
| Gene Symbol: Bik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053704.1 | NP_446156.1 | 4 | 444 | ||
| XM_006242048.2 | XP_006242110.1 | 4 | 531 | |||
| GenBank mRNA | AF372501.1 | - | 4 | 444 | ||
| FQ225338.1 | - | 4 | 421 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bik| Gene Symbol: Bik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053704.1 | NP_446156.1 | 4 | 444 | ||
| XM_006242048.2 | XP_006242110.1 | 4 | 531 | |||
| GenBank mRNA | AF372501.1 | - | 4 | 444 | ||
| FQ225338.1 | - | 4 | 421 | |||