siRNA Details
| Gene Symbol: Lmx1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234024.2 | XP_006234086.1 | 4 | 766 | ||
| GenBank mRNA | AF390073.1 | - | 1 | 18 | ||
| AY169317.1 | - | 2 | 262 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lmx1b| Gene Symbol: Lmx1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006234024.2 | XP_006234086.1 | 4 | 766 | ||
| GenBank mRNA | AF390073.1 | - | 1 | 18 | ||
| AY169317.1 | - | 2 | 262 | |||