siRNA Details
| Gene Symbol: Klf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053713.1 | NP_446165.1 | 4 | 1334 | ||
| GenBank mRNA | AF390546.1 | - | 4 | 1334 | ||
| BC085720.1 | - | 4 | 1727 | 7190219 | ||
| FQ233252.1 | - | 2 | 701 | |||
| FQ233720.1 | - | 2 | 702 | |||

Klf4
Genomic Map