siRNA Details
| Gene Symbol: Ankh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053714.1 | NP_446166.1 | 6 | 1114 | ||
| GenBank mRNA | AF393241.1 | - | 6 | 1114 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankh| Gene Symbol: Ankh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053714.1 | NP_446166.1 | 6 | 1114 | ||
| GenBank mRNA | AF393241.1 | - | 6 | 1114 | ||