siRNA Details
Gene Symbol: Slc5a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053715.2 | NP_446167.2 | 1 | 993 | ||
GenBank mRNA | AJ001290.1 | - | 1 | 993 | ||
AY231162.1 | - | 1 | 934 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc5a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053715.2 | NP_446167.2 | 1 | 993 | ||
GenBank mRNA | AJ001290.1 | - | 1 | 993 | ||
AY231162.1 | - | 1 | 934 |