siRNA Details
| Gene Symbol: Emb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053719.1 | NP_446171.1 | 6 | 707 | ||
| GenBank mRNA | AJ009698.1 | - | 6 | 707 | ||
| BC061846.1 | - | 6 | 709 | 5621074 | ||
| FQ213962.1 | - | 6 | 720 | |||
| FQ228768.1 | - | 6 | 720 | |||

Emb
Genomic Map