siRNA Details
| Gene Symbol: Lancl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053723.1 | NP_446175.1 | 7, 8 | 1190 | ||
| GenBank mRNA | AJ131111.1 | - | 7, 8 | 1190 | ||
| BC085811.1 | - | 7, 8 | 1168 | 7115423 | ||
| FQ211687.1 | - | 2, 3 | 295 | |||

Lancl1
Genomic Map