siRNA Details
| Gene Symbol: Glra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053724.3 | NP_446176.3 | 644 | |||
| GenBank mRNA | AJ310838.1 | - | 644 | |||
| M55250.1 | - | 995 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Glra3| Gene Symbol: Glra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053724.3 | NP_446176.3 | 644 | |||
| GenBank mRNA | AJ310838.1 | - | 644 | |||
| M55250.1 | - | 995 | ||||